Assay Details
Target Gene Details
Entrez Gene ID: | 1063 |
Gene Name: | centromere protein F |
Gene Aliases: |
CENF, CILD31, PRO1779, STROMS, hcp-1 |
Location: |
Chr.1:214603181-214664574 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPF | NM_016343.3 | NP_057427.3 | ||
| XM_011509082.2 | XP_011507384.1 | |||
| XM_017000086.1 | XP_016855575.1 | |||
| AK307690.1 | ||||
| BU662228.1 | ||||
| U30872.1 | AAA82935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv574n100 | Chr.1:214590435 - 214840032 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map