Assay Details
Target Gene Details
Entrez Gene ID: | 3065 |
Gene Name: | histone deacetylase 1 |
Gene Aliases: |
GON-10, HD1, RPD3, RPD3L1 |
Location: |
Chr.1:32292103-32333628 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC1 | NM_004964.2 | NP_004955.2 | ||
| XM_011541309.2 | XP_011539611.1 | |||
| AB451297.1 | ||||
| AB451430.1 | ||||
| AK225555.1 | ||||
| AK296308.1 | ||||
| AK299241.1 | ||||
| AK299792.1 | ||||
| AK313061.1 | ||||
| AY627042.1 | AAT44863.1 | |||
| BC000301.2 | AAH00301.1 | |||
| CR541717.1 | CAG46518.1 | |||
| D50405.1 | BAA08909.1 | |||
| U50079.1 | AAC50475.1 |
More Information
Set Membership: |
|

Genomic Map