Assay Details
Target Gene Details
Entrez Gene ID: | 2948 |
Gene Name: | glutathione S-transferase mu 4 |
Gene Aliases: |
GSTM4-4, GTM4 |
Location: |
Chr.1:109656076-109665501 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTM4 | NM_147148.2 | NP_671489.1 | ||
| AI949679.1 | ||||
| BI791533.1 | ||||
| M99422.1 | AAA58623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv258n100 | Chr.1:109658958 - 109700319 on Build GRCh38 | Gain |
|
| dgv257n100 | Chr.1:109650144 - 109707602 on Build GRCh38 | Gain |
|
| dgv254n100 | Chr.1:109631914 - 109700319 on Build GRCh38 | Gain+Loss |
|
| dgv77e212 | Chr.1:109635559 - 109709330 on Build GRCh38 | Loss |
|
| nsv547486 | Chr.1:109657738 - 109690459 on Build GRCh38 | Loss |
|
| dgv256n100 | Chr.1:109638683 - 109700319 on Build GRCh38 | Loss |
|
| nsv2299 | Chr.1:109610296 - 109662341 on Build GRCh38 | Deletion |
|
| esv25172 | Chr.1:109644224 - 109716486 on Build GRCh38 | Gain+Loss |
|
| esv3587153 | Chr.1:109660894 - 109674476 on Build GRCh38 | Loss |
|
| dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map