Assay Details
Target Gene Details
Entrez Gene ID: | 23507 |
Gene Name: | leucine rich repeat containing 8 family member B |
Gene Aliases: |
TA-LRRP, TALRRP |
Location: |
Chr.1:89522627-89597861 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC8B | NM_001134476.1 | NP_001127948.1 | ||
| XM_005270701.4 | XP_005270758.1 | |||
| XM_005270702.4 | XP_005270759.1 | |||
| XM_005270703.4 | XP_005270760.1 | |||
| XM_011541143.2 | XP_011539445.1 | |||
| XM_011541144.2 | XP_011539446.1 | |||
| XM_011541145.2 | XP_011539447.1 | |||
| XM_011541146.2 | XP_011539448.1 | |||
| XM_011541148.2 | XP_011539450.1 | |||
| XM_017000885.1 | XP_016856374.1 | |||
| XM_017000886.1 | XP_016856375.1 | |||
| BC030607.2 | AAH30607.1 | |||
| BC060782.1 | AAH60782.1 | |||
| BI459217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586765 | Chr.1:89419740 - 89647558 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72957878] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map