Assay Details
Target Gene Details
Entrez Gene ID: | 92241 |
Gene Name: | RCSD domain containing 1 |
Gene Aliases: |
CAPZIP, MK2S4 |
Location: |
Chr.1:167630193-167708696 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCSD1 | NM_001322923.1 | NP_001309852.1 | ||
| NM_001322924.1 | NP_001309853.1 | |||
| NM_052862.3 | NP_443094.3 | |||
| NR_136519.1 | ||||
| AF545852.1 | AAN52359.1 | |||
| AK130798.1 | ||||
| AL137762.1 | CAB70910.1 | |||
| BC101536.1 | AAI01537.1 | |||
| BC101562.1 | AAI01563.1 | |||
| BC143428.1 | ||||
| DR006615.1 |
More Information
Set Membership: |
|

Genomic Map