Assay Details
Target Gene Details
Entrez Gene ID: | 94236 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5 |
Gene Aliases: |
HEJ1 |
Location: |
Chr.1:101872012-101894743 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA1P5 | NR_033424.2 | |||
| AF395440.1 | AAK73122.1 |
Target Gene Details
Entrez Gene ID: | 118427 |
Gene Name: | olfactomedin 3 |
Gene Aliases: |
NOE3, NOELIN3, OPTIMEDIN |
Location: |
Chr.1:101800028-101997234 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLFM3 | NM_001288823.1 | NP_001275752.1 | ||
| NM_058170.3 | NP_477518.2 | |||
| NR_110210.1 | ||||
| NR_110211.1 | ||||
| AF397394.1 | AAK97475.1 | |||
| AF397395.1 | AAK97476.1 | |||
| AF397396.1 | AAK97477.1 | |||
| AF397397.1 | AAK97478.1 | |||
| AK095724.1 | ||||
| BC022531.1 | AAH22531.1 | |||
| DA406512.1 | ||||
| DA528521.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522156 | Chr.1:101844093 - 101956158 on Build GRCh38 | Gain |
|
| esv3586982 | Chr.1:101884648 - 101892953 on Build GRCh38 | Loss |
|
| esv3582582 | Chr.1:101843693 - 101958966 on Build GRCh38 | Gain |
|
| esv3582471 | Chr.1:101578331 - 102371530 on Build GRCh38 | Gain |
|
| nsv522921 | Chr.1:101704739 - 102410836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map