Assay Details
Target Gene Details
Entrez Gene ID: | 84432 |
Gene Name: | prokineticin 1 |
Gene Aliases: |
EGVEGF, PK1, PRK1 |
Location: |
Chr.1:110451166-110457354 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROK1 | NM_032414.2 | NP_115790.1 | ||
| AF333024.1 | AAK49918.1 | |||
| AY029225.1 | AAK33111.1 | |||
| AY358683.1 | AAQ89046.1 | |||
| BC025399.1 | AAH25399.1 | |||
| BM544315.1 |
More Information
Set Membership: |
|

Genomic Map