Assay Details
Target Gene Details
Entrez Gene ID: | 10100 |
Gene Name: | tetraspanin 2 |
Gene Aliases: |
NET3, TSN2, TSPAN-2 |
Location: |
Chr.1:115048011-115089501 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN2 | NM_001308315.1 | NP_001295244.1 | ||
| NM_001308316.1 | NP_001295245.1 | |||
| NM_005725.5 | NP_005716.2 | |||
| XM_016999996.1 | XP_016855485.1 | |||
| AF054839.1 | AAC69715.1 | |||
| AK315385.1 | ||||
| BC021675.1 | AAH21675.1 | |||
| BQ067895.1 | ||||
| CB993308.1 | ||||
| GU971730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26121 | Chr.1:115020681 - 115560070 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map