Assay Details
Target Gene Details
Entrez Gene ID: | 57554 |
Gene Name: | leucine rich repeat containing 7 |
Gene Aliases: |
DENSIN |
Location: |
Chr.1:69567130-70152426 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC7 | XM_017001885.1 | XP_016857374.1 | ||
| XM_017001886.1 | XP_016857375.1 | |||
| XM_017001887.1 | XP_016857376.1 | |||
| XM_017001888.1 | XP_016857377.1 | |||
| XM_017001889.1 | XP_016857378.1 | |||
| XM_017001890.1 | XP_016857379.1 | |||
| XM_017001891.1 | XP_016857380.1 | |||
| XM_017001892.1 | XP_016857381.1 | |||
| XM_017001893.1 | XP_016857382.1 | |||
| XM_017001894.1 | XP_016857383.1 | |||
| XM_017001895.1 | XP_016857384.1 | |||
| AF498274.1 | AAP06801.1 | |||
| AK294206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830126 | Chr.1:69549196 - 69702296 on Build GRCh38 | Gain |
|
| esv3577911 | Chr.1:69582653 - 69594225 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map