Assay Details
Target Gene Details
Entrez Gene ID: | 400799 |
Gene Name: | long intergenic non-protein coding RNA 1344 |
Gene Aliases: |
GS1-122H1.2 |
Location: |
Chr.1:182203945-182314061 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01344 | NR_104175.1 | |||
| AK128015.1 | ||||
| BX096536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588097 | Chr.1:182203948 - 182218948 on Build GRCh38 | Loss |
|
| esv2658469 | Chr.1:182203950 - 182218951 on Build GRCh38 | Deletion |
|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|

Genomic Map