Assay Details
Target Gene Details
Entrez Gene ID: | 400794 |
Gene Name: | uncharacterized LOC400794 |
Gene Aliases: |
- |
Location: |
Chr.1:165476842-165582155 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400794 | NR_026744.2 | |||
| AK093132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3376 | Chr.1:165513885 - 165548586 on Build GRCh38 | Insertion |
|

Genomic Map