Assay Details
Target Gene Details
Entrez Gene ID: | 100861504 |
Gene Name: | long intergenic non-protein coding RNA 538 |
Gene Aliases: |
PROX1UT, Yiya |
Location: |
Chr.1:213924749-213926654 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00538 | NR_046189.1 | 1 | 931 | |
| JF495099.1 | 1 | 931 |
Target Gene Details
Entrez Gene ID: | 100505832 |
Gene Name: | PROX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:213819641-213986153 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROX1-AS1 | NR_037850.2 | |||
| AK092251.1 | ||||
| BE179719.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139810890,rs76881878] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map