Assay Details
Target Gene Details
Entrez Gene ID: | 51094 |
Gene Name: | adiponectin receptor 1 |
Gene Aliases: |
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A |
Location: |
Chr.1:202940825-202958572 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADIPOR1 | NM_001290629.1 | NP_001277558.1 | ||
| BX461217.2 |
More Information
Set Membership: |
|

Genomic Map