Assay Details
Target Gene Details
Entrez Gene ID: | 25782 |
Gene Name: | RAB3 GTPase activating non-catalytic protein subunit 2 |
Gene Aliases: |
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150 |
Location: |
Chr.1:220148268-220272501 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3GAP2 | NM_012414.3 | NP_036546.2 | ||
| AB020646.2 | BAA74862.2 | |||
| AF004828.1 | AAC35881.1 | |||
| AF255648.1 | AAG44636.1 | |||
| AK021928.1 | BAB13939.1 | |||
| AK027485.1 | ||||
| AK291234.1 | ||||
| AL117631.1 | CAB56022.1 | |||
| BC146760.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79503392] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map