Assay Details
Target Gene Details
Entrez Gene ID: | 4259 |
Gene Name: | microsomal glutathione S-transferase 3 |
Gene Aliases: |
GST-III |
Location: |
Chr.1:165630873-165656136 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST3 | NM_004528.3 | NP_004519.1 | ||
| XM_005245174.3 | XP_005245231.2 | |||
| AK312103.1 | ||||
| BC000505.2 | AAH00505.1 | |||
| BC005964.1 | AAH05964.1 | |||
| BI666647.1 | ||||
| DQ185045.1 | ABD14425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548096 | Chr.1:165634629 - 165635552 on Build GRCh38 | Loss |
|
| esv28160 | Chr.1:165634326 - 165635211 on Build GRCh38 | Loss |
|
| nsv548090 | Chr.1:165633287 - 165635754 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map