Assay Details
Target Gene Details
Entrez Gene ID: | 81573 |
Gene Name: | ankyrin repeat domain 13C |
Gene Aliases: |
dJ677H15.3 |
Location: |
Chr.1:70259002-70354977 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13C | NM_030816.4 | NP_110443.3 | ||
| XM_005271234.3 | XP_005271291.1 | |||
| XM_005271235.3 | XP_005271292.1 | |||
| XM_006710929.3 | XP_006710992.1 | |||
| XM_017002413.1 | XP_016857902.1 | |||
| XM_017002414.1 | XP_016857903.1 | |||
| XM_017002415.1 | XP_016857904.1 | |||
| XM_017002416.1 | XP_016857905.1 | |||
| AK074187.1 | ||||
| AL136717.1 | CAB66651.1 | |||
| BC028840.1 | AAH28840.1 | |||
| BU622247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014292 | Chr.1:70223532 - 70333382 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75774314] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map