Assay Details
Target Gene Details
Entrez Gene ID: | 2275 |
Gene Name: | four and a half LIM domains 3 |
Gene Aliases: |
SLIM2 |
Location: |
Chr.1:37996765-38005515 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL3 | NM_001243878.1 | NP_001230807.1 | ||
| NM_004468.4 | NP_004459.2 | |||
| AK290641.1 | ||||
| AK309179.1 | ||||
| BC001351.2 | AAH01351.1 | |||
| BC011697.2 | AAH11697.1 | |||
| BC014501.1 | AAH14501.1 | |||
| BX384389.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs76026335] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map