Assay Details
Target Gene Details
Entrez Gene ID: | 8574 |
Gene Name: | aldo-keto reductase family 7 member A2 |
Gene Aliases: |
AFAR, AFAR1, AFB1-AR1, AKR7 |
Location: |
Chr.1:19302708-19312146 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR7A2 | NM_001320979.1 | NP_001307908.1 | ||
| NM_003689.3 | NP_003680.2 | |||
| AF026947.1 | AAC52104.1 | |||
| AK225267.1 | ||||
| AK303131.1 | ||||
| BC004111.2 | AAH04111.3 | |||
| BC007352.2 | AAH07352.2 | |||
| BC010852.1 | AAH10852.1 | |||
| BC011586.1 | AAH11586.1 | |||
| BC012171.1 | AAH12171.1 | |||
| BC013996.2 | AAH13996.1 | |||
| BT007347.1 | AAP36011.1 | |||
| BX401279.2 | ||||
| EU794591.1 | ||||
| Y16675.1 | CAA76347.1 |
More Information
Set Membership: |
|

Genomic Map