Assay Details
Target Gene Details
Entrez Gene ID: | 5690 |
Gene Name: | proteasome subunit beta 2 |
Gene Aliases: |
HC7-I |
Location: |
Chr.1:35599541-35641844 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMB2 | NM_001199779.1 | NP_001186708.1 | ||
| NM_001199780.1 | NP_001186709.1 | |||
| NM_002794.4 | NP_002785.1 | |||
| AB209470.1 | BAD92707.1 | |||
| AI025755.1 | ||||
| AK296956.1 | ||||
| AK310023.1 | ||||
| AV762678.2 | ||||
| BC101836.1 | AAI01837.1 | |||
| BC105126.1 | AAI05127.1 | |||
| BC107901.1 | AAI07902.1 | |||
| BM849239.1 | ||||
| BT007137.1 | AAP35801.1 | |||
| CR456862.1 | CAG33143.1 | |||
| D26599.1 | BAA05646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012556 | Chr.1:35413704 - 35703660 on Build GRCh38 | Gain |
|
| esv3585723 | Chr.1:35627175 - 35641806 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114197012] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map