Assay Details
Target Gene Details
Entrez Gene ID: | 128372 |
Gene Name: | olfactory receptor family 6 subfamily N member 1 |
Gene Aliases: |
OR1-22 |
Location: |
Chr.1:158747733-158840979 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6N1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv591n54 | Chr.1:158745498 - 158759556 on Build GRCh38 | Loss |
|
| dgv589n54 | Chr.1:158715246 - 158883163 on Build GRCh38 | Loss |
|
| nsv824920 | Chr.1:158756287 - 158758335 on Build GRCh38 | Loss |
|
| dgv56e214 | Chr.1:158724216 - 158757728 on Build GRCh38 | Loss |
|
| dgv592n54 | Chr.1:158755555 - 158758153 on Build GRCh38 | Loss |
|
| esv2562592 | Chr.1:158755528 - 158756479 on Build GRCh38 | Insertion |
|
| nsv1002401 | Chr.1:158742837 - 159002241 on Build GRCh38 | Loss |
|

Genomic Map