Assay Details
Target Gene Details
Entrez Gene ID: | 57795 |
Gene Name: | BMP/retinoic acid inducible neural specific 2 |
Gene Aliases: |
DBCCR1L2, FAM5B |
Location: |
Chr.1:177170922-177282422 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRINP2 | XM_005245379.1 | XP_005245436.1 | ||
| AK296524.1 |
More Information
Set Membership: |
|

Genomic Map