Assay Details
Target Gene Details
Entrez Gene ID: | 10262 |
Gene Name: | splicing factor 3b subunit 4 |
Gene Aliases: |
AFD1, Hsh49, SAP49, SF3b49 |
Location: |
Chr.1:149923317-149928252 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B4 | NM_005850.4 | NP_005841.1 | ||
| AK097315.1 | ||||
| AK223322.1 | BAD97042.1 | |||
| AK310495.1 | ||||
| BC004273.1 | AAH04273.1 | |||
| BC013886.2 | AAH13886.1 | |||
| BC090883.1 | AAH90883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831537 | Chr.1:149861791 - 150092561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map