Assay Details
Target Gene Details
Entrez Gene ID: | 54558 |
Gene Name: | spermatogenesis associated 6 |
Gene Aliases: |
HASH, SRF-1, SRF1 |
Location: |
Chr.1:48258643-48472208 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA6 | XM_006710699.2 | XP_006710762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585965 | Chr.1:48282463 - 48313854 on Build GRCh38 | Loss |
|
| esv2762139 | Chr.1:48279401 - 48310271 on Build GRCh38 | Loss |
|
| nsv1011588 | Chr.1:48248029 - 48328196 on Build GRCh38 | Gain |
|
| esv3578249 | Chr.1:48202315 - 48332128 on Build GRCh38 | Gain |
|
| nsv508182 | Chr.1:48264259 - 48353795 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72887987] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map