Assay Details
Target Gene Details
Entrez Gene ID: | 3776 |
Gene Name: | potassium two pore domain channel subfamily K member 2 |
Gene Aliases: |
K2p2.1, TPKC1, TREK, TREK-1, TREK1, hTREK-1c, hTREK-1e |
Location: |
Chr.1:215002920-215237093 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK2 | NM_001017424.2 | NP_001017424.1 | ||
| XM_017001248.1 | XP_016856737.1 | |||
| XM_017001249.1 | XP_016856738.1 | |||
| AK123409.1 | BAC85609.1 | |||
| AU280191.1 | ||||
| AY552980.2 | ||||
| AY552981.1 | AAT49016.1 | |||
| CD625291.1 | ||||
| CD625297.1 | ||||
| DA475072.1 | ||||
| KF182338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549182 | Chr.1:214993893 - 215075074 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79884578] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map