Assay Details
Target Gene Details
Entrez Gene ID: | 58480 |
Gene Name: | ras homolog family member U |
Gene Aliases: |
ARHU, CDC42L1, G28K, WRCH1, hG28K |
Location: |
Chr.1:228644647-228746669 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOU | NR_037962.1 | |||
| DA321957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891181 | Chr.1:228670108 - 228853454 on Build GRCh38 | Gain |
|
| nsv998255 | Chr.1:228656037 - 228783455 on Build GRCh38 | Gain |
|
| nsv549299 | Chr.1:228657499 - 229320129 on Build GRCh38 | Gain |
|
| nsv549300 | Chr.1:228718956 - 228783455 on Build GRCh38 | Loss |
|
| esv3589057 | Chr.1:228711090 - 228720005 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map