Assay Details
Target Gene Details
Entrez Gene ID: | 22977 |
Gene Name: | aldo-keto reductase family 7 member A3 |
Gene Aliases: |
AFAR2 |
Location: |
Chr.1:19274309-19289362 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR7A3 | XM_011541046.2 | 7 | 1643 | XP_011539348.1 |
| XM_017000714.1 | 7 | 801 | XP_016856203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3552788 | Chr.1:19266487 - 19285352 on Build GRCh38 | Deletion |
|
| esv26443 | Chr.1:19273138 - 19288033 on Build GRCh38 | Loss |
|
| dgv25e199 | Chr.1:19270091 - 19287665 on Build GRCh38 | Deletion |
|
| esv3585392 | Chr.1:19272870 - 19287742 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148806318] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map