Assay Details
Target Gene Details
Entrez Gene ID: | 8543 |
Gene Name: | LIM domain only 4 |
Gene Aliases: |
- |
Location: |
Chr.1:87328468-87348923 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO4 | NM_006769.3 | 5 | 2740 | NP_006760.1 |
| XM_005271291.3 | 5 | 4910 | XP_005271348.1 |
More Information
Set Membership: |
|

Genomic Map