Assay Details
Target Gene Details
Entrez Gene ID: | 5629 |
Gene Name: | prospero homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.1:213983239-214041510 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROX1 | NM_001270616.1 | NP_001257545.1 | ||
| XM_005273194.1 | XP_005273251.1 | |||
| XM_011509771.1 | XP_011508073.1 | |||
| XM_011509773.2 | XP_011508075.1 | |||
| AK298182.1 | ||||
| DA802102.1 | ||||
| DB086260.1 | ||||
| U44060.1 | AAC50656.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952088 | Chr.1:213980058 - 213991157 on Build GRCh38 | Deletion |
|
| nsv4498 | Chr.1:213981463 - 214016219 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map