Assay Details
Target Gene Details
Entrez Gene ID: | 7139 |
Gene Name: | troponin T2, cardiac type |
Gene Aliases: |
CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT |
Location: |
Chr.1:201359008-201377828 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470776 | Chr.1:201319827 - 201534604 on Build GRCh38 | Gain |
|
| nsv467961 | Chr.1:201316584 - 201534604 on Build GRCh38 | Gain |
|
| nsv947466 | Chr.1:201308515 - 201540498 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map