Assay Details
Target Gene Details
Entrez Gene ID: | 10163 |
Gene Name: | WAS protein family member 2 |
Gene Aliases: |
IMD2, SCAR2, WASF4, WAVE2, dJ393P12.2 |
Location: |
Chr.1:27404226-27490187 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WASF2 | NM_001201404.2 | NP_001188333.1 | ||
| NM_006990.4 | NP_008921.1 | |||
| AK303011.1 | ||||
| BC032724.1 | ||||
| BC040943.1 | AAH40943.1 | |||
| DC300388.1 | ||||
| DC376823.1 | ||||
| HY001891.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77902868] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map