Assay Details
Target Gene Details
Entrez Gene ID: | 23612 |
Gene Name: | pleckstrin homology like domain family A member 3 |
Gene Aliases: |
TIH1 |
Location: |
Chr.1:201464284-201469171 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLDA3 | NM_012396.4 | 2 | 2440 | NP_036528.1 |
| AI083980.1 | ||||
| AI188803.1 | 1 | 198 | ||
| AI624156.1 | 1 | 295 | ||
| AI678238.1 | ||||
| AI870979.1 | 1 | 74 | ||
| AW194162.1 | ||||
| AW513769.1 | ||||
| BE778799.1 | ||||
| BE892338.1 | 1 | 581 | ||
| BG990425.1 | ||||
| BM996951.1 | 1 | 88 | ||
| BQ000725.1 | ||||
| CA406398.1 | 1 | 240 | ||
| N94465.1 | ||||
| W30744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832315 | Chr.1:201364463 - 201525988 on Build GRCh38 | Loss |
|
| nsv470776 | Chr.1:201319827 - 201534604 on Build GRCh38 | Gain |
|
| nsv467961 | Chr.1:201316584 - 201534604 on Build GRCh38 | Gain |
|
| nsv832326 | Chr.1:201375720 - 201475887 on Build GRCh38 | Loss |
|
| nsv947466 | Chr.1:201308515 - 201540498 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map