Assay Details
Target Gene Details
Entrez Gene ID: | 54482 |
Gene Name: | tRNA methyltransferase 13 homolog |
Gene Aliases: |
CCDC76 |
Location: |
Chr.1:100133080-100150498 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT13 | NM_019083.2 | NP_061956.2 | ||
| NR_135078.1 | ||||
| XM_005270945.3 | XP_005271002.1 | |||
| XM_005270946.2 | XP_005271003.1 | |||
| XM_017001504.1 | XP_016856993.1 | |||
| XM_017001505.1 | XP_016856994.1 | |||
| AK001149.1 | BAA91520.1 | |||
| AK002081.1 | BAA92074.1 | |||
| AK289404.1 | ||||
| AK294738.1 | ||||
| AK295828.1 | ||||
| AK298941.1 | ||||
| AL699548.1 | ||||
| BC075811.1 | AAH75811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2121 | Chr.1:100115063 - 100149647 on Build GRCh38 | Insertion |
|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map