Assay Details
Target Gene Details
Entrez Gene ID: | 100507178 |
Gene Name: | SLFNL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:41014590-41043890 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLFNL1-AS1 | NR_037868.1 | 1 | 317 | |
| AL157461.1 | 1 | 317 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546103 | Chr.1:41011772 - 41076832 on Build GRCh38 | Loss |
|

Genomic Map