Assay Details
Target Gene Details
Entrez Gene ID: | 22826 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C8 |
Gene Aliases: |
HSPC331, SPF31 |
Location: |
Chr.1:28200278-28233031 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC8 | NM_014280.2 | NP_055095.2 | ||
| AF083190.1 | AAC35352.1 | |||
| AF161433.1 | AAF28993.1 | |||
| AF161449.1 | AAF29009.1 | |||
| AK300797.1 | ||||
| BC033159.1 | AAH33159.1 | |||
| BX538122.1 | ||||
| CR456900.1 | CAG33181.1 | |||
| DB294321.1 |
More Information
Set Membership: |
|

Genomic Map