Assay Details
Target Gene Details
Entrez Gene ID: | 400823 |
Gene Name: | family with sequence similarity 177 member B |
Gene Aliases: |
- |
Location: |
Chr.1:222737216-222772193 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM177B | XM_017001279.1 | 5 | 2989 | XP_016856768.1 |
| XM_017001280.1 | 6 | 2503 | XP_016856769.1 | |
| XM_017001281.1 | 6 | 3072 | XP_016856770.1 | |
| XM_017001282.1 | 6 | 2528 | XP_016856771.1 | |
| XM_017001285.1 | XP_016856774.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116384086] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map