Assay Details
Target Gene Details
Entrez Gene ID: | 2035 |
Gene Name: | erythrocyte membrane protein band 4.1 |
Gene Aliases: |
4.1R, EL1, HE |
Location: |
Chr.1:28887091-29120046 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
|

Genomic Map