Assay Details
Target Gene Details
Entrez Gene ID: | 257101 |
Gene Name: | zinc finger protein 683 |
Gene Aliases: |
Hobit |
Location: |
Chr.1:26361632-26374520 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF683 | XM_005245828.3 | 1 | 185 | XP_005245885.1 |
| XM_006710555.3 | 1 | 185 | XP_006710618.1 | |
| XM_017000954.1 | 1 | 185 | XP_016856443.1 | |
| XM_017000957.1 | 1 | 185 | XP_016856446.1 | |
| XM_017000958.1 | 1 | 185 | XP_016856447.1 | |
| XM_017000959.1 | 1 | 185 | XP_016856448.1 |
More Information
Set Membership: |
|

Genomic Map