Assay Details
Target Gene Details
Entrez Gene ID: | 55706 |
Gene Name: | NDC1 transmembrane nucleoporin |
Gene Aliases: |
NET3, TMEM48 |
Location: |
Chr.1:53765460-53838552 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDC1 | NM_001168551.1 | NP_001162023.1 | ||
| NM_018087.4 | NP_060557.3 | |||
| NR_033142.1 | ||||
| XM_011541766.2 | XP_011540068.1 | |||
| AK001269.1 | BAA91592.1 | |||
| AK022618.1 | BAB14135.1 | |||
| AK091439.1 | BAC03665.1 | |||
| AK295000.1 | ||||
| AK298909.1 | ||||
| AK302910.1 | ||||
| AL354612.1 | CAB89725.1 | |||
| AL354613.1 | CAB89726.1 | |||
| BC003082.2 | AAH03082.1 | |||
| BX648031.1 | ||||
| DQ141696.1 | AAZ73087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011262 | Chr.1:53542956 - 54106304 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map