Assay Details
Target Gene Details
Entrez Gene ID: | 5906 |
Gene Name: | RAP1A, member of RAS oncogene family |
Gene Aliases: |
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21 |
Location: |
Chr.1:111542223-111716695 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP1A | XM_017001964.1 | XP_016857453.1 | ||
| M22995.1 | AAA36150.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75905417] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map