Assay Details
Target Gene Details
Entrez Gene ID: | 5351 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 |
Gene Aliases: |
EDS6, LH, LH1, LLH, PLOD |
Location: |
Chr.1:11934667-11975542 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLOD1 | NM_000302.3 | NP_000293.2 | ||
| NM_001316320.1 | NP_001303249.1 | |||
| AK294688.1 | ||||
| AK295254.1 | ||||
| AK299150.1 | ||||
| AK312243.1 | ||||
| AK316285.1 | ||||
| AU139472.1 | ||||
| BC016657.2 | AAH16657.1 | |||
| DC334246.1 | ||||
| M98252.1 |
More Information
Set Membership: |
|

Genomic Map