Assay Details
Target Gene Details
Entrez Gene ID: | 55631 |
Gene Name: | leucine rich repeat containing 40 |
Gene Aliases: |
dJ677H15.1 |
Location: |
Chr.1:70144802-70205678 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC40 | NM_017768.4 | NP_060238.3 | ||
| XM_005271013.1 | XP_005271070.1 | |||
| XM_011541763.1 | XP_011540065.1 | |||
| XM_017001695.1 | XP_016857184.1 | |||
| XM_017001696.1 | XP_016857185.1 | |||
| AK000338.1 | BAA91094.1 | |||
| AK022951.1 | BAB14326.1 | |||
| AK225337.1 | ||||
| AL162076.1 | CAB82410.1 | |||
| BC003407.1 | AAH03407.2 | |||
| BC008586.1 | AAH08586.1 |
Target Gene Details
Entrez Gene ID: | 57554 |
Gene Name: | leucine rich repeat containing 7 |
Gene Aliases: |
DENSIN |
Location: |
Chr.1:69567130-70152426 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC7 | XM_017001886.1 | XP_016857375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005397 | Chr.1:70133120 - 70168902 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map