Assay Details
Target Gene Details
Entrez Gene ID: | 100505832 |
Gene Name: | PROX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:213819641-213986153 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROX1-AS1 | NR_037850.2 | |||
| AK092251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275485 | Chr.1:213830375 - 213840793 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79443654] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map