Assay Details
Target Gene Details
Entrez Gene ID: | 22977 |
Gene Name: | aldo-keto reductase family 7 member A3 |
Gene Aliases: |
AFAR2 |
Location: |
Chr.1:19274309-19289362 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR7A3 | NM_012067.2 | NP_036199.2 | ||
| XM_011541046.2 | XP_011539348.1 | |||
| AF040639.1 | AAD02195.1 | |||
| AJ271799.1 | CAC81076.1 | |||
| BC025709.1 | AAH25709.1 | |||
| BC031562.1 | AAH31562.1 | |||
| BC042420.1 | ||||
| BM855386.1 | ||||
| BM920315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26443 | Chr.1:19273138 - 19288033 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map