Assay Details
Target Gene Details
Entrez Gene ID: | 118427 |
Gene Name: | olfactomedin 3 |
Gene Aliases: |
NOE3, NOELIN3, OPTIMEDIN |
Location: |
Chr.1:101800028-101997234 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLFM3 | NM_001288821.1 | NP_001275750.1 | ||
| NM_001288823.1 | NP_001275752.1 | |||
| NM_058170.3 | NP_477518.2 | |||
| NR_110210.1 | ||||
| NR_110211.1 | ||||
| NR_110212.1 | ||||
| XM_017000240.1 | XP_016855729.1 | |||
| AF397392.2 | AAK97473.1 | |||
| AF397393.2 | AAK97474.1 | |||
| AF397394.1 | AAK97475.1 | |||
| AF397395.1 | AAK97476.1 | |||
| AF397396.1 | AAK97477.1 | |||
| AF397397.1 | AAK97478.1 | |||
| AK095551.1 | ||||
| AK095724.1 | ||||
| AK300870.1 | ||||
| AY358722.1 | AAQ89084.1 | |||
| BC022531.1 | AAH22531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582471 | Chr.1:101578331 - 102371530 on Build GRCh38 | Gain |
|
| dgv379n54 | Chr.1:101808720 - 101838596 on Build GRCh38 | Loss |
|
| nsv2143 | Chr.1:101788623 - 101823251 on Build GRCh38 | Insertion |
|
| nsv522921 | Chr.1:101704739 - 102410836 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77546134] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map