Assay Details
Target Gene Details
Entrez Gene ID: | 55699 |
Gene Name: | isoleucyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
CAGSSS, ILERS |
Location: |
Chr.1:220094113-220148041 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IARS2 | NM_018060.3 | NP_060530.3 | ||
| AK001188.1 | BAA91544.1 | |||
| AK001726.1 | ||||
| AK022665.1 | BAB14164.1 | |||
| AK291432.1 | ||||
| AY267462.1 | ||||
| BC010218.1 | AAH10218.2 | |||
| BC040376.1 | ||||
| BC047880.1 | AAH47880.3 | |||
| BC137438.1 | ||||
| D28500.1 | BAA95147.1 |
Target Gene Details
Entrez Gene ID: | 406969 |
Gene Name: | microRNA 194-1 |
Gene Aliases: |
MIRN194-1, mir-194-1 |
Location: |
Chr.1:220118157-220118241 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR194-1 | NR_029711.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832636 | Chr.1:219966355 - 220121010 on Build GRCh38 | Loss |
|
| nsv947468 | Chr.1:220114634 - 220119407 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map