Assay Details
Target Gene Details
Entrez Gene ID: | 27101 |
Gene Name: | calcyclin binding protein |
Gene Aliases: |
GIG5, PNAS-107, S100A6BP, SIP |
Location: |
Chr.1:174999435-175012027 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACYBP | NM_001007214.1 | NP_001007215.1 | ||
| NM_014412.2 | NP_055227.1 | |||
| XM_017001046.1 | XP_016856535.1 | |||
| AF057356.1 | AAC21458.1 | |||
| AF275803.1 | AAG23817.1 | |||
| AF314752.1 | AAG34170.1 | |||
| AK093425.1 | ||||
| AK313278.1 | ||||
| AL035305.1 | CAA22910.1 | |||
| AY423723.1 | AAS00486.1 | |||
| BC005975.1 | AAH05975.1 | |||
| BC022352.1 | AAH22352.1 | |||
| BC067823.1 | AAH67823.1 | |||
| BC078151.1 | AAH78151.1 | |||
| BQ432793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001000 | Chr.1:174975071 - 175009717 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78442918] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map