Assay Details
Target Gene Details
Entrez Gene ID: | 9015 |
Gene Name: | TATA-box binding protein associated factor, RNA polymerase I subunit A |
Gene Aliases: |
MGC:17061, RAFI48, SL1, TAFI48 |
Location: |
Chr.1:222557902-222590096 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF1A | NM_001201536.1 | NP_001188465.1 | ||
| NM_005681.3 | NP_005672.1 | |||
| NM_139352.2 | NP_647603.1 | |||
| XM_005273343.3 | XP_005273400.1 | |||
| XM_006711612.1 | XP_006711675.1 | |||
| XM_006711613.3 | XP_006711676.1 | |||
| XM_017002758.1 | XP_016858247.1 | |||
| XM_017002759.1 | XP_016858248.1 | |||
| XM_017002760.1 | XP_016858249.1 | |||
| AK001054.1 | BAA91482.1 | |||
| AK290970.1 | ||||
| AK299536.1 | ||||
| AK315740.1 | ||||
| BC013808.1 | AAH13808.1 | |||
| L39060.1 | AAA62862.1 |
More Information
Set Membership: |
|

Genomic Map