Assay Details
Target Gene Details
Entrez Gene ID: | 336 |
Gene Name: | apolipoprotein A2 |
Gene Aliases: |
Apo-AII, ApoA-II, apoAII |
Location: |
Chr.1:161222293-161223628 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOA2 | NM_001643.1 | NP_001634.1 | ||
| AB106565.1 | BAD05173.1 | |||
| AK312034.1 | ||||
| BC005282.1 | AAH05282.1 | |||
| BT006786.1 | AAP35432.1 | |||
| M29882.1 | AAA51701.1 | |||
| X00955.1 | CAA25467.1 |
More Information
Set Membership: |
|

Genomic Map