Assay Details
Target Gene Details
Entrez Gene ID: | 1382 |
Gene Name: | cellular retinoic acid binding protein 2 |
Gene Aliases: |
CRABP-II, RBP6 |
Location: |
Chr.1:156699606-156712900 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRABP2 | NM_001199723.1 | NP_001186652.1 | ||
| NM_001878.3 | NP_001869.1 | |||
| XM_017000343.1 | XP_016855832.1 | |||
| AB593017.1 | ||||
| AK312007.1 | ||||
| AU126559.1 | ||||
| BC001109.2 | AAH01109.1 | |||
| BP231157.1 | ||||
| BT019827.1 | AAV38630.1 | |||
| CR450357.1 | CAG29353.1 | |||
| M68867.1 | AAA52068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map