Assay Details
Target Gene Details
Entrez Gene ID: | 10485 |
Gene Name: | chromosome 1 open reading frame 61 |
Gene Aliases: |
CROC4 |
Location: |
Chr.1:156404252-156456649 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf61 | XM_011509067.1 | XP_011507369.1 | ||
| XM_011509068.1 | XP_011507370.1 | |||
| XM_017000061.1 | XP_016855550.1 | |||
| XM_017000062.1 | XP_016855551.1 | |||
| XM_017000064.1 | XP_016855553.1 | |||
| XM_017000066.1 | XP_016855555.1 |

Genomic Map